Learn More
JDM LAB SOLUTIONS LLC Benchtop Mini centrifuge with speed of 6000RPM and 9200 RPM
Catalog No. 502558505
Encompass
Benchtop Mini centrifuge with speed of 6000RPM and 9200 RPM;
Included accessories fit a variety of tubes, with a capacity of 8 x 1.5mL/2.0mL Tubes (8 x 0.5mL with supplied adapters) / 2 x 0.2mL Tube Strips or 16 x 0.2mL Tubes / 4 x 5mL Tubes
Catalog No. 50-255-8505 Supplier JDM LAB SOLUTIONS LLC Supplier No. MLX200FA
Benchtop Mini centrifuge with speed of 6000RPM and 9200 RPM;
Included accessories fit a variety of tubes, with a capacity of 8 x 1.5mL/2.0mL Tubes (8 x 0.5mL with supplied adapters) / 2 x 0.2mL Tube Strips or 16 x 0.2mL Tubes / 4 x 5mL Tubes