Learn More
Ace Glass, Inc. Benchtop Optical Polariscope
SDP
Supplier:  Ace Glass, Inc. 12024110
Encompass
Benchtop optical polariscope designed for inspection of internal stress patterns in glass quartz and transparent plastic materials. Unit utilizes LED illumination to provide bright uniform light for clear visualization of birefringence and strain. Compact design with durable aluminum housing allows for easy placement in laboratory or production environments. Ideal for quality control inspection material testing and glass fabrication applications. Simple operation with no calibration required ensures reliable repeatable results. Suitable for routine analysis educational use and manufacturing inspection processes.
Catalog No. 50-008-54680