Learn More
Ace Glass, Inc. Benchtop Optical Polariscope
SDP
Catalog No. 5000854831
Encompass
Benchtop optical polariscope designed for inspection of internal stress patterns in glass quartz and transparent plastic materials. Unit utilizes LED illumination to provide bright uniform light for clear visualization of birefringence and strain. Compact design with durable aluminum housing allows for easy placement in laboratory or production environments. Ideal for quality control inspection material testing and glass fabrication applications. Simple operation with no calibration required ensures reliable repeatable results. Suitable for routine analysis educational use and manufacturing inspection processes.
Catalog No. 50-008-54831 Supplier Ace Glass, Inc. Supplier No. 12024220
Benchtop optical polariscope designed for inspection of internal stress patterns in glass quartz and transparent plastic materials. Unit utilizes LED illumination to provide bright uniform light for clear visualization of birefringence and strain. Compact design with durable aluminum housing allows for easy placement in laboratory or production environments. Ideal for quality control inspection material testing and glass fabrication applications. Simple operation with no calibration required ensures reliable repeatable results. Suitable for routine analysis educational use and manufacturing inspection processes.