Learn More
Azzota Corp Benchtop pH meter -1-15pH mV
Catalog No. 5000887303
Encompass_Preferred
BENCHTOP PH METER WITH MV/ C/ F PH range -1 00 15 00pH mV range 1999mV
Catalog No. 50-008-87303 Supplier Azzota Corp Supplier No. PHMV210
BENCHTOP PH METER WITH MV/ C/ F PH range -1 00 15 00pH mV range 1999mV