Learn More
Azzota Corp Benchtop Ph/Mv Meter With Temp. Ph Range: 0.00-14.00Ph Ph Accuracy: +/-0.01Ph Mv Range: 0 To +/-1999Mv Mv Accuracy: +/-2Mv Temperature Range: 0 To 100 Cdeg.
Catalog No. 507532967
Encompass_Preferred
Benchtop pH/mV Meter with Temp. pH Range: 0.00-14.00pH pH Accuracy: +/-0.01pH mV Range: 0 to +/-1999mV mV Accuracy: +/-2mV Temperature Range: 0 to 100 Cdeg.
Catalog No. 50-753-2967 Supplier Azzota Corp Supplier No. PHMV1T
Benchtop pH/mV Meter with Temp. pH Range: 0.00-14.00pH pH Accuracy: +/-0.01pH mV Range: 0 to +/-1999mV mV Accuracy: +/-2mV Temperature Range: 0 to 100 Cdeg.