Learn More
Chemglass Life Sciences Benchtop Support Stand, Mobile, Assembly Supplied with the Following: Reactor Support Frame, Single & Dual Base Plates, Support Rods, Cork Rings, 250mL Adapter Plate, and 300mL-1000mL Adapter Plate 
SDP
Catalog No. CG1951100
Encompass_Preferred
Benchtop Support Stand Features:• 3/4" Stainless Steel Pipe (1.050" OD) Uprights and Heavy Duty PTFE Coated Aluminum Fittings Provide Maximum Strength and Chemical Durability• All Stainless Steel Uprights are Passivated to Inhibit Corrosion• Adjustable Stainless Steel Shelf/Tray• Supports Reactors from 250mL to 5L: Single Setup is for Reactor Sizes 300mL to 5L; Dual Setup is for Reactor Sizes 250 to 1000mL• Middle Support Bracket is Vertically Adjustable and is Supplied with Hook and Loop Clamps for Reactors up to 1000mL and a Set of Heavy Duty Clamps for use with 2 thru 5L Reactors• Top Support Bracket is Vertically Adjustable and Provides for Attachment of a Single or Two Overhead Stirrers. Mounting Clamps Accept Overhead Stirrers with a MAX Support Arm of 5/8” Diameter• Unique Design Allows Easy Installation/Removal of Glassware• Single and Dual Reactor Support Plates have Built-In Bubble Levels• Mobile: Large Swivel Casters with Locking Mechanisms • 19” W x 19” D x 71.25” H
Catalog No. CG1951100 Supplier Chemglass Life Sciences Supplier No. CG1951100
Benchtop Support Stand Features:• 3/4" Stainless Steel Pipe (1.050" OD) Uprights and Heavy Duty PTFE Coated Aluminum Fittings Provide Maximum Strength and Chemical Durability• All Stainless Steel Uprights are Passivated to Inhibit Corrosion• Adjustable Stainless Steel Shelf/Tray• Supports Reactors from 250mL to 5L: Single Setup is for Reactor Sizes 300mL to 5L; Dual Setup is for Reactor Sizes 250 to 1000mL• Middle Support Bracket is Vertically Adjustable and is Supplied with Hook and Loop Clamps for Reactors up to 1000mL and a Set of Heavy Duty Clamps for use with 2 thru 5L Reactors• Top Support Bracket is Vertically Adjustable and Provides for Attachment of a Single or Two Overhead Stirrers. Mounting Clamps Accept Overhead Stirrers with a MAX Support Arm of 5/8” Diameter• Unique Design Allows Easy Installation/Removal of Glassware• Single and Dual Reactor Support Plates have Built-In Bubble Levels• Mobile: Large Swivel Casters with Locking Mechanisms • 19” W x 19” D x 71.25” H