Learn More
PCE AMERICAS INC Analytical Balance scale with high-precision counting with a measuring range of up to 220 g.
SDP
Catalog No. NC3654596
Encompass_Preferred
"The benchtop scale is a high-precision benchtop scale with a measuring range of up to 220 g. Particularly noteworthy about the benchtop scale is the large LCD touch screen. Among other things, the benchtop scale can still piece counting, percent weighing and density determination.- Measuring range: 0 ... 220 g- Resolution: 0.1 mg- Measurement accuracy: ± 0.5 mg- 5"" touch screen- Windbreak- RS232 data interface"
Catalog No. NC3654596 Supplier PCE AMERICAS INC Supplier No. PCEABT 220
"The benchtop scale is a high-precision benchtop scale with a measuring range of up to 220 g. Particularly noteworthy about the benchtop scale is the large LCD touch screen. Among other things, the benchtop scale can still piece counting, percent weighing and density determination.- Measuring range: 0 ... 220 g- Resolution: 0.1 mg- Measurement accuracy: ± 0.5 mg- 5"" touch screen- Windbreak- RS232 data interface"