Learn More
Poltex Benchtop Simple Riser Shelf with Drawer
SDP
Catalog No. 21377567
Catalog No. 21-377-567 Supplier Poltex Supplier No. RISERBIN1288
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
Simple benchtop riser shelf with a drawer underneath.
  • Simple benchtop shelf with drawer underneath
  • Super durable 1/4 in. thick PETG construction
  • Rubber bumpons on base for non-slip placement
  • 12 x 8 x 8 in.
Specifications
Product Type Riser
Autoclavable Not Autoclavable
Dimensions (L x W x H) 12 x 8 x 8 in.
With Casters No