Learn More
Andwin Scientific Yamato 18L Steam Sterilizer, Benchtop, 220V
SDP
Catalog No. 501898059
Encompass_Preferred
Space-saving, low cost 18L benchtop steam sterilizer, ideal for research facilities. Easy to use. Powerful 1500W pipe heater. Easy to read 4 digit LED display. Programmable sterilizing and temperature functions. Timer Setting Range 0 to 999 min. Increased safety features: Water level detection sensor with alarm, Overheat Protection, Sensor Lid closure sensor (Inter lock), Pressure Lamp Indicator, New improved silicon lid gasket which is easy to replace. Temp setting range 50 to 126°C
Catalog No. 50-189-8059 Supplier Andwin Scientific Supplier No. SK111C
Space-saving, low cost 18L benchtop steam sterilizer, ideal for research facilities. Easy to use. Powerful 1500W pipe heater. Easy to read 4 digit LED display. Programmable sterilizing and temperature functions. Timer Setting Range 0 to 999 min. Increased safety features: Water level detection sensor with alarm, Overheat Protection, Sensor Lid closure sensor (Inter lock), Pressure Lamp Indicator, New improved silicon lid gasket which is easy to replace. Temp setting range 50 to 126°C