Learn More
Research Products International Corp BenchWaver 3D Rocker with flat mat
SDP
Catalog No. 502229816
Encompass
The BenchWaver combines the basic rocking motion of traditional platform rockers with the circular motion of an orbital shaker. This combination is ideal for generating the optimum 3-dimensional “undulating” motion for molecular biology applications. BenchWaver is a reliable and versatile instrument and can be used In refrigerated or incubated environments from 2° to 60°C.
The tilt angle isinstantly adjustable from 0 to10° and the mixing speed can be adjusted from 5 to 105 rpm, using the large, user-friendly LCD control panel. Combined control of both parameters (tilt and speed) makes the BenchWaver a versatile workhorse that can provide anything from gentle, slow moving waves to a rapid, vigorous motion. Additionally, timer runs from 1 minute, up to 99 hours, or continuous operation. The large13.5 x13 inch platform is supplied complete with a non-slip, rubber mat.
Catalog No. 50-222-9816 Supplier Research Products International Corp Supplier No. 260133
The BenchWaver combines the basic rocking motion of traditional platform rockers with the circular motion of an orbital shaker. This combination is ideal for generating the optimum 3-dimensional “undulating” motion for molecular biology applications. BenchWaver is a reliable and versatile instrument and can be used In refrigerated or incubated environments from 2° to 60°C.
The tilt angle isinstantly adjustable from 0 to10° and the mixing speed can be adjusted from 5 to 105 rpm, using the large, user-friendly LCD control panel. Combined control of both parameters (tilt and speed) makes the BenchWaver a versatile workhorse that can provide anything from gentle, slow moving waves to a rapid, vigorous motion. Additionally, timer runs from 1 minute, up to 99 hours, or continuous operation. The large13.5 x13 inch platform is supplied complete with a non-slip, rubber mat.