Learn More
Aviva Systems Biology Corp Bend6 Antibody - C-terminal region (ARP55514_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55514_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Bend6. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: RGD1310392, Bend6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-047