Learn More
Aviva Systems Biology Corp BEND7 Antibody - middle region (ARP55524_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55524_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C10orf30. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BEND7. Alternate names: FLJ40283, MGC35247, C10orf30ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54018