Learn More
Cayman Chemical BendamustIn-d4hydrochlorId 1mg
SDP
Catalog No. 502991117
Encompass_Preferred
An internal standard for the quantification of bendamustine
Catalog No. 50-299-1117 Supplier Cayman Chemical Supplier No. 313161 MG
Add to Cart
Add to Cart
An internal standard for the quantification of bendamustine