Learn More
Medchemexpress LLC Bendamustine | 16506-27-7 | MFCD00866481 | 98.0% | C16H21Cl2N3O2 | 10MG
SDP
Catalog No. 5000258577
Encompass_Preferred
Bendamustine is a research-grade analytical standard of the purine analogue SDX-105 free base (CAS 16506-27-7) used as a DNA cross-linking agent with alkylating and antimetabolite activity. Supplied as a solid with defined purity and storage guidance, it is intended for analytical and research applications such as mechanism-of-action studies and assay development.
  • High purity (98.0%) suitable for analytical workflows.
  • Chemical identity supported by SDS and spectral data.
  • Solid form; store at -20°C and protect from light.
  • Appropriate for DNA damage and apoptosis research.
  • Available in small analytical pack sizes for assay development.
Catalog No. 50-002-58577 Supplier Medchemexpress LLC Supplier No. HY1356710MG
Bendamustine is a research-grade analytical standard of the purine analogue SDX-105 free base (CAS 16506-27-7) used as a DNA cross-linking agent with alkylating and antimetabolite activity. Supplied as a solid with defined purity and storage guidance, it is intended for analytical and research applications such as mechanism-of-action studies and assay development.
  • High purity (98.0%) suitable for analytical workflows.
  • Chemical identity supported by SDS and spectral data.
  • Solid form; store at -20°C and protect from light.
  • Appropriate for DNA damage and apoptosis research.
  • Available in small analytical pack sizes for assay development.