Learn More
Medchemexpress LLC Bendamustine (free base) | 16506-27-7 | MFCD00866481 | 98.0% | 358.26 g/mol | C16H21Cl2N3O2 | 5 MG
SDP
Catalog No. 5000221987
Encompass_Preferred
Bendamustine (SDX-105 free base) is a purine analogue alkylating agent that induces DNA cross-linking, activating DNA-damage responses and apoptosis. Supplied for research use, it is used in studies of cytotoxicity, apoptosis induction, and lymphoma or leukemia models.
  • Functions as a DNA cross-linking alkylating agent.
  • Activates DNA-damage response and apoptosis.
  • Suitable for cytotoxicity and cell-death pathway research.
  • Provided as the free base form.
  • Reported purity 98.0%.
  • CAS 16506-27-7; molecular weight 358.26 g/mol.
Catalog No. 50-002-21987 Supplier Medchemexpress LLC Supplier No. HY135675MG
Bendamustine (SDX-105 free base) is a purine analogue alkylating agent that induces DNA cross-linking, activating DNA-damage responses and apoptosis. Supplied for research use, it is used in studies of cytotoxicity, apoptosis induction, and lymphoma or leukemia models.
  • Functions as a DNA cross-linking alkylating agent.
  • Activates DNA-damage response and apoptosis.
  • Suitable for cytotoxicity and cell-death pathway research.
  • Provided as the free base form.
  • Reported purity 98.0%.
  • CAS 16506-27-7; molecular weight 358.26 g/mol.