Learn More
Selleck Chemical LLC Bendamustine HCl 100mg 3543-75-7 SDX-105 (Cytostasane) HCl
SDP
Catalog No. 507013
Encompass_Preferred
Bendamustine (SDX-105, Cytostasane) HCl is a DNA-damaging agent with IC50 of 50 ?M in cell-free assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-701-3 Supplier Selleck Chemical LLC Supplier No. S1212100MG
Bendamustine (SDX-105, Cytostasane) HCl is a DNA-damaging agent with IC50 of 50 ?M in cell-free assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.