Learn More
Medchemexpress LLC Bendamustine (hydrochloride) | 3543-75-7 | 99.89% | C16H22Cl3N3O2 | 100 MG
SDP
Catalog No. 5000432542
Encompass_Preferred
Bendamustine hydrochloride is a purine analogue and a DNA cross-linking agent that activates DNA-damage stress response and apoptosis. It possesses potent alkylating, anticancer, and antimetabolite properties.
  • A purine analogue.
  • Acts as a DNA cross-linking agent.
  • Activates DNA-damage stress response.
  • Induces apoptosis.
  • Exhibits potent alkylating properties.
  • Demonstrates anticancer effects.
  • Possesses antimetabolite characteristics.
  • Induces p21 (Cip1/Waf1) and NOXA genes.
  • Increases the expression of p53 in SU-DHL-1 cells.
  • Blocks mitotic checkpoints and causes mitotic catastrophe.
  • Reduces the viability of multiple myeloma cell lines.
  • Induces specific caspase-dependent multiple myeloma cell death.
  • Inhibits the spindle-assembly checkpoint.
Catalog No. 50-004-32542 Supplier Medchemexpress LLC Supplier No. HYB0077100MG
Bendamustine hydrochloride is a purine analogue and a DNA cross-linking agent that activates DNA-damage stress response and apoptosis. It possesses potent alkylating, anticancer, and antimetabolite properties.
  • A purine analogue.
  • Acts as a DNA cross-linking agent.
  • Activates DNA-damage stress response.
  • Induces apoptosis.
  • Exhibits potent alkylating properties.
  • Demonstrates anticancer effects.
  • Possesses antimetabolite characteristics.
  • Induces p21 (Cip1/Waf1) and NOXA genes.
  • Increases the expression of p53 in SU-DHL-1 cells.
  • Blocks mitotic checkpoints and causes mitotic catastrophe.
  • Reduces the viability of multiple myeloma cell lines.
  • Induces specific caspase-dependent multiple myeloma cell death.
  • Inhibits the spindle-assembly checkpoint.