Learn More
TARGETMOL CHEMICALS INC Bendamustine hydrochloride
SDP
Catalog No. 502880645
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Bendamustine hydrochloride (EP-3101) (IC50 of 50 uM) is an alkylating agent associated with DNA damage. Purity 98.03%
Catalog No. 50-288-0645 Supplier TARGETMOL CHEMICALS INC Supplier No. T0095100MG
Also available in 1 mL, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Bendamustine hydrochloride (EP-3101) (IC50 of 50 uM) is an alkylating agent associated with DNA damage. Purity 98.03%