Learn More
Abcam Bendamustine hydrochloride, DNA alkylating agent, 10MG
Catalog No. 502706908
Encompass_Preferred
MW 394.7 Da, Purity >99%. DNA alkylating agent. Causes DNA intrastrand and interstrand cross-links. Induces apoptotic gene expression. Anticancer, apoptotic agent in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6908 Supplier Abcam Supplier No. AB14109610MG
MW 394.7 Da, Purity >99%. DNA alkylating agent. Causes DNA intrastrand and interstrand cross-links. Induces apoptotic gene expression. Anticancer, apoptotic agent in vivo.
The product is subject to the following: Abcam Restricted Use Statement