Learn More
Medchemexpress LLC Bendazac | 20187-55-7 | MFCD00866158 | 99.43% | 282.29 | 1 ML
SDP
Catalog No. 5000386893
Encompass_Preferred
Bendazac is an oxyacetic acid that possesses anti-inflammatory, antinecrotic, choleretic, and antilipidaemic properties. It functions by preventing protein denaturation and also contributes to delaying the cataractogenic process.
  • Anti-inflammatory properties
  • Antinecrotic properties
  • Choleretic properties
  • Antilipidaemic properties
  • Prevents protein denaturation
  • Delays the cataractogenic process
Catalog No. 50-003-86893 Supplier Medchemexpress LLC Supplier No. HY1748010MM/1ML
Bendazac is an oxyacetic acid that possesses anti-inflammatory, antinecrotic, choleretic, and antilipidaemic properties. It functions by preventing protein denaturation and also contributes to delaying the cataractogenic process.
  • Anti-inflammatory properties
  • Antinecrotic properties
  • Choleretic properties
  • Antilipidaemic properties
  • Prevents protein denaturation
  • Delays the cataractogenic process