Learn More
Selleck Chemical LLC Bendazac S5361-25mg
SDP
Catalog No. 503109970
Encompass_Preferred
Bendazac (bendazolic acid AF-983) is a non-steroidal anti-inflammatory drug that delays the cataractogenic process It may act by preventing protein denaturation
Catalog No. 50-310-9970 Supplier Selleck Chemical LLC Supplier No. S536125MG
Bendazac (bendazolic acid AF-983) is a non-steroidal anti-inflammatory drug that delays the cataractogenic process It may act by preventing protein denaturation