Learn More
Medchemexpress LLC Bendroflumethiazide-d5 | 1330183-13-5 | 98.6% | 1 MG
SDP
Catalog No. 5000412855
Encompass_Preferred
Bendroflumethiazide-d5 is a deuterium-labeled version of Bendroflumethiazide, an orally available diuretic. It functions by inhibiting the electroneutral sodium-chloride symporter in the distal convoluted tubule, effectively lowering blood pressure. This compound is valuable in the study of hypertension and edema, and demonstrates an antidiuretic effect in diabetes insipidus.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-12855 Supplier Medchemexpress LLC Supplier No. HYB1363S1MG
Bendroflumethiazide-d5 is a deuterium-labeled version of Bendroflumethiazide, an orally available diuretic. It functions by inhibiting the electroneutral sodium-chloride symporter in the distal convoluted tubule, effectively lowering blood pressure. This compound is valuable in the study of hypertension and edema, and demonstrates an antidiuretic effect in diabetes insipidus.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.