Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benedict's Qualitative Solution, Innovating Science™
SCI_ED

Catalog No. S000534 Shop All Aldon Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3.8 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000534 1 L
S000533 500 mL
S000535 3.8 L
S000536 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Warning: Harmful if swallowed, may be harmful in contact with skin, causes mild skin irritation, causes eye irritation, harmful if inhaled, very toxic to aquatic life with long lasting effects.
Catalog No. S000534 Supplier Aldon Corporation Supplier No. IS11033
Add to Cart
Add to Cart
Warning: Harmful if swallowed, may be harmful in contact with skin, causes mild skin irritation, causes eye irritation, harmful if inhaled, very toxic to aquatic life with long lasting effects.

Detect sugars in substances.

  • Green = Trace Sugars
  • Yellow = Low Sugars
  • Orange = Moderate Sugars
  • Red/Brown = High Sugars

Specifications

CAS 7732-18-5, 68-04-2, 497-19-8, 7758-99-8
Grade Reagent
Packaging HDPE
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.