Learn More
Medchemexpress LLC Benfluorex (hydrochloride) | 23642-66-2 | 99.6% | 1 G
SDP
Catalog No. 5000413864
Encompass_Preferred
Benfluorex hydrochloride (JP-992 hydrochloride) is a hepatic nuclear factor 4 alpha (HNF4α) activator. This compound is for research use only and activates insulin promoter activity.
  • Increases GFP-positive cells
  • Increases endogenous insulin mRNA
  • Stimulates HNF4α expression
  • Alters HNF4α protease sensitivity
  • Decreases synthesis of acid-soluble products and ketone bodies from oleate
  • Increases 14CO2 production into the citric acid cycle
  • Inhibits gluconeogenesis from lactate/pyruvate
Catalog No. 50-004-13864 Supplier Medchemexpress LLC Supplier No. HYB10581G
Benfluorex hydrochloride (JP-992 hydrochloride) is a hepatic nuclear factor 4 alpha (HNF4α) activator. This compound is for research use only and activates insulin promoter activity.
  • Increases GFP-positive cells
  • Increases endogenous insulin mRNA
  • Stimulates HNF4α expression
  • Alters HNF4α protease sensitivity
  • Decreases synthesis of acid-soluble products and ketone bodies from oleate
  • Increases 14CO2 production into the citric acid cycle
  • Inhibits gluconeogenesis from lactate/pyruvate