Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benfluorex hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111013636 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3636 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3636 Supplier MilliporeSigma Supelco Supplier No. B75225G
Add to Cart
Add to Cart

Benfluorex hydrochloride salt is a hypolipidaemic compound with lipid and glucose lowering effects.It is used in the treatment of impaired carbohydrate metabolism, atherogenic metabolic disorders and in the suppression of appetite.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Poly Bottle
CAS 23642-66-2
Physical Form Neat
Linear Formula C19H20F3 NO2 · HCl
MDL Number MFCD00058493
Synonym N-(1-Methyl-2-(3-[trifluoromethyl]-phenyl)ethyl]aminoethanol benzoate ester
Type Analytical Standard
Quantity 5 g
Formula Weight 387.82
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.