Learn More
Medchemexpress LLC HY-B2045 50mg Medchemexpress, Benfluralin CAS:1861-40-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB204550MG
Encompass_Preferred
Medchemexpress, HY-B2045 50mg Benfluralin CAS:1861-40-1 Benfluralin is a kind of herbicide and an agrochemical which can be used as a pre-emergence herbicide used for the control of grass and other weeds in a range of food and non-food crops. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3051