Learn More
Apexbio Technology LLC Benidipine HCl 91599-74-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000091197
Encompass_Preferred
Benidipine hydrochloride (CAS 91599-74-5) is a dihydropyridine-class small molecule that functions as an inhibitor of voltage-dependent calcium channels thereby modulating calcium influx in vascular smooth muscle and cardiac tissue Additionally benidipine hydrochloride has been shown to suppress mineralocorticoid receptor coactivator (MRC) activation induced by aldosterone This dual inhibitory activity makes it valuable for biomedical research into cardiovascular physiology calcium signaling pathways and the molecular mechanisms underlying mineralocorticoid receptor-associated conditions
Catalog No. 50-000-91197 Supplier Apexbio Technology LLC Supplier No. B14095.1
Benidipine hydrochloride (CAS 91599-74-5) is a dihydropyridine-class small molecule that functions as an inhibitor of voltage-dependent calcium channels thereby modulating calcium influx in vascular smooth muscle and cardiac tissue Additionally benidipine hydrochloride has been shown to suppress mineralocorticoid receptor coactivator (MRC) activation induced by aldosterone This dual inhibitory activity makes it valuable for biomedical research into cardiovascular physiology calcium signaling pathways and the molecular mechanisms underlying mineralocorticoid receptor-associated conditions