Learn More
Cayman Chemical BenIdIpInhydrochlorIde 100mg
SDP
Catalog No. 502964227
Encompass_Preferred
An orally bioavailable inhibitor of L-, T-, and N-type calcium channels; IC50 = 2.7 nM in guinea pig ventricular cells; prevents oxidative stress in vitro and in neural stem cells; has antihypertensive effects; competitive antagonist at mineralocorticoid receptors
Catalog No. 50-296-4227 Supplier Cayman Chemical Supplier No. 21607100 MG
An orally bioavailable inhibitor of L-, T-, and N-type calcium channels; IC50 = 2.7 nM in guinea pig ventricular cells; prevents oxidative stress in vitro and in neural stem cells; has antihypertensive effects; competitive antagonist at mineralocorticoid receptors