Learn More
Cayman Chemical Benocyclidine, Purity: ≥98%, CAS: 112726-66-6, 50 mg
SDP
Catalog No. 502954823
Encompass_Preferred
A derivative of phencyclidine that acts as a potent and selective dopamine reuptake inhibitor (IC50 = 8 nM); intended for forensic and research applications
Catalog No. 50-295-4823 Supplier Cayman Chemical Supplier No. 1173750 MG
A derivative of phencyclidine that acts as a potent and selective dopamine reuptake inhibitor (IC50 = 8 nM); intended for forensic and research applications