Learn More
Chem Service Inc Benomyl,17804-35-2,100MG
SDP
Supplier:  Chem Service Inc N11138100MG
Encompass_Preferred
Molecular Formula C14H18N4O3, Purity 99.50%, Molecular Weight 290.32, Store in cool dry place, Appearance: Colorless to white, crestalline solid, Synonyms: Methyl-1-[butylcarbamoyl]-2-benzimidazole Carbamate
Catalog No. NC9651088