Learn More
Medchemexpress LLC HY-107795 500mg Medchemexpress, Benorilate CAS:5003-48-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY107795500MG
Encompass_Preferred
Medchemexpress, HY-107795 500mg Benorilate CAS:5003-48-5 Benorylate (Salipran) is the esterification product of paracetamol and acetylsalicylic acid. Benorylate has anti-inflammatory, analgesic and antipyretic properties. Benorylate could also inhibit prostaglandin (PG) synthesis[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0720