Learn More
Cayman Chemical Benoxaprofen 10mg
SDP
Catalog No. 502912982
Encompass_Preferred
An NSAID; inhibits PGE2 and LTC4 production in A23187-stimulated isolated mouse peritoneal macrophages at 100 µM; inhibits serum-induced chemotaxis of isolated rat peritoneal leukocytes at 25 and 50 UG/ml; induces hemolysis in isolated human erythrocytes and histamine release in isolated rat peritoneal mast cells in a light-dependent manner at 10 µM; reduces carrageenan-induced paw edema and yeast-induced fever in rats and acetic acid-induced writhing in mice
Catalog No. 50-291-2982 Supplier Cayman Chemical Supplier No. 3600110 MG
An NSAID; inhibits PGE2 and LTC4 production in A23187-stimulated isolated mouse peritoneal macrophages at 100 µM; inhibits serum-induced chemotaxis of isolated rat peritoneal leukocytes at 25 and 50 UG/ml; induces hemolysis in isolated human erythrocytes and histamine release in isolated rat peritoneal mast cells in a light-dependent manner at 10 µM; reduces carrageenan-induced paw edema and yeast-induced fever in rats and acetic acid-induced writhing in mice