Learn More
Medchemexpress LLC Benpyrine | 2550398-89-3 | MFCD32015709 | 99.3% | C16H16N6O | 10MG
SDP
Catalog No. 5000259935
Encompass_Preferred
Benpyrine is a small-molecule TNF-α inhibitor supplied for laboratory research use. It is available as a solid and as a 10 mM solution in DMSO, and is characterized by high purity, a defined molecular formula, and a moderate molecular weight.
  • High purity (≈99.3%).
  • Available as solid in multiple pack sizes (5-100 mg) and as 10 mM solution in 1 mL DMSO.
  • Molecular formula C16H16N6O, molecular weight 308.34.
  • Suitable for biochemical and cellular TNF-α studies.
  • Store powder at -20°C for long-term stability.
Catalog No. 50-002-59935 Supplier Medchemexpress LLC Supplier No. HY13380710MG
Benpyrine is a small-molecule TNF-α inhibitor supplied for laboratory research use. It is available as a solid and as a 10 mM solution in DMSO, and is characterized by high purity, a defined molecular formula, and a moderate molecular weight.
  • High purity (≈99.3%).
  • Available as solid in multiple pack sizes (5-100 mg) and as 10 mM solution in 1 mL DMSO.
  • Molecular formula C16H16N6O, molecular weight 308.34.
  • Suitable for biochemical and cellular TNF-α studies.
  • Store powder at -20°C for long-term stability.