Learn More
Apexbio Technology LLC Benserazide HCl 14919-77-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000088751
Encompass_Preferred
Benserazide HCl is a small-molecule inhibitor targeting aromatic L-amino acid decarboxylase (AADC) It is designed to inhibit peripheral AADC activity thereby reducing dopamine synthesis outside the central nervous system and facilitating increased availability of administered levodopa to the brain Benserazide HCl exerts its biological activity primarily through selective inhibition of peripheral AADC In experimental research it is frequently combined with levodopa to investigate dopamine metabolism and neural transmission pathways Based on these pharmacological properties Benserazide HCl holds research potential in studies involving neurodegenerative disease models particularly Parkinson s disease to explore dopaminergic system dynamics and therapeutic mechanisms
Catalog No. 50-000-88751 Supplier Apexbio Technology LLC Supplier No. B14895.1
Benserazide HCl is a small-molecule inhibitor targeting aromatic L-amino acid decarboxylase (AADC) It is designed to inhibit peripheral AADC activity thereby reducing dopamine synthesis outside the central nervous system and facilitating increased availability of administered levodopa to the brain Benserazide HCl exerts its biological activity primarily through selective inhibition of peripheral AADC In experimental research it is frequently combined with levodopa to investigate dopamine metabolism and neural transmission pathways Based on these pharmacological properties Benserazide HCl holds research potential in studies involving neurodegenerative disease models particularly Parkinson s disease to explore dopaminergic system dynamics and therapeutic mechanisms