Learn More
Enzo Life Sciences Benserazide. HCl (1g). CAS: 14919-77-8
SDP
Supplier:  Enzo Life Sciences BMLEI1651000
Encompass_Preferred
Peripheral decarboxylase inhibitor. Also inhibits tryptophan hydroxylase and tyrosine hydroxylase. Used with L-DOPA as a Parkinson’s disease therapeutic. Purity: ≥98%. Solubility: Soluble in water. Almost insoluble in 100% ethanol, insoluble in acetone. Long Term Storage: Ambient.
Catalog No. 50-201-0583