Learn More
Medchemexpress LLC Benserazide (hydrochloride) | 14919-77-8 | MFCD00078571 | 99.7% | 1 G
SDP
Catalog No. 5000405294
Encompass_Preferred
Benserazide hydrochloride is an aromatic L-amino acid decarboxylase (AADC) and L-DOPA decarboxylase inhibitor. It is also a PKM2 inhibitor that directly binds to and blocks PKM2 enzyme activity, leading to the inhibition of aerobic glycolysis and concurrent up-regulation of OXPHOS. It can be used for the study of Parkinson's disease and melanoma.
  • Aromatic L-amino acid decarboxylase (AADC) and L-DOPA decarboxylase inhibitor
  • PKM2 inhibitor
  • Directly binds to and blocks PKM2 enzyme activity
  • Inhibits aerobic glycolysis
  • Up-regulates OXPHOS
  • Useful for the study of Parkinson's disease and melanoma
Catalog No. 50-004-05294 Supplier Medchemexpress LLC Supplier No. HYB0404A1G
Benserazide hydrochloride is an aromatic L-amino acid decarboxylase (AADC) and L-DOPA decarboxylase inhibitor. It is also a PKM2 inhibitor that directly binds to and blocks PKM2 enzyme activity, leading to the inhibition of aerobic glycolysis and concurrent up-regulation of OXPHOS. It can be used for the study of Parkinson's disease and melanoma.
  • Aromatic L-amino acid decarboxylase (AADC) and L-DOPA decarboxylase inhibitor
  • PKM2 inhibitor
  • Directly binds to and blocks PKM2 enzyme activity
  • Inhibits aerobic glycolysis
  • Up-regulates OXPHOS
  • Useful for the study of Parkinson's disease and melanoma