Learn More
Selleck Chemical LLC Benserazide HCl 10mg 14919-77-8 Ro-4-4602
SDP
Catalog No. 505071
Encompass_Preferred
Benserazide HCl (Ro-4-4602) is a peripherally-acting aromatic L-amino acid decarboxylase (AAAD) or DOPA decarboxylase inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-507-1 Supplier Selleck Chemical LLC Supplier No. S245310MG
Benserazide HCl (Ro-4-4602) is a peripherally-acting aromatic L-amino acid decarboxylase (AAAD) or DOPA decarboxylase inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.