Learn More
Cayman Chemical BensRazIDd3hydroCLOrID 2.5mg
SDP
Catalog No. 502990557
Encompass_Preferred
An internal standard for the quantification of benserazide by GC- or LC-MS
Catalog No. 50-299-0557 Supplier Cayman Chemical Supplier No. 296432.5 MG
An internal standard for the quantification of benserazide by GC- or LC-MS