Learn More
Medchemexpress LLC Bensulfuron-methyl | 83055-99-6 | 98.4% | 25 MG
SDP
Catalog No. 5000450458
Encompass_Preferred
Bensulfuron-methyl is a sulfonylurea herbicide that is widely used to control broad-leaf weeds in rice paddies.
  • Sulfonylurea herbicide
  • Widely used to control broad-leaf weeds
  • Used in rice paddies
Catalog No. 50-004-50458 Supplier Medchemexpress LLC Supplier No. 25MGHYB0870
Bensulfuron-methyl is a sulfonylurea herbicide that is widely used to control broad-leaf weeds in rice paddies.
  • Sulfonylurea herbicide
  • Widely used to control broad-leaf weeds
  • Used in rice paddies