Learn More
Selleck Chemical LLC Bentamapimod S8867-2MG
SDP
Catalog No. 5000024500
Encompass_Preferred
Bentamapimod (AS602801) is a novel orally active JNK inhibitor with IC50 values of 80 nM 90 nM and 230 nM for JNK1 JNK2 and JNK3 respectively
Catalog No. 50-000-24500 Supplier Selleck Chemical LLC Supplier No. S88672MG
Bentamapimod (AS602801) is a novel orally active JNK inhibitor with IC50 values of 80 nM 90 nM and 230 nM for JNK1 JNK2 and JNK3 respectively