Learn More
Medchemexpress LLC Bentazone-d7 | 131842-77-8 | MFCD09953755 | 98.73% | 1 MG
SDP
Catalog No. 5000410571
Encompass_Preferred
Bentazone-d7 is the deuterium labeled Bentazone. Bentazone is a post-emergence herbicide used for selective control of broadleaf weeds and sedges in beans, rice, corn, peanuts, mint and others. It acts by interfering with photosynthesis.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-10571 Supplier Medchemexpress LLC Supplier No. HYB2039S11MG
Bentazone-d7 is the deuterium labeled Bentazone. Bentazone is a post-emergence herbicide used for selective control of broadleaf weeds and sedges in beans, rice, corn, peanuts, mint and others. It acts by interfering with photosynthesis.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.