Learn More
Medchemexpress LLC HY-B1493 5mg Medchemexpress, Bentiromide CAS:37106-97-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB14935MG
Encompass_Preferred
Medchemexpress, HY-B1493 5mg Bentiromide CAS:37106-97-1 Bentiromide is a peptide that is broken down in the pancreas by chymotrypsin. The bentiromide test is an excellent means of confirming the diagnosis of pancreatic exocrine insufficiency by outpatient test of chymotrypsin function [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2954