Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bentonite, MP Biomedicals

Catalog No. MP021008945 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021008945 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021008945 Supplier MP Biomedicals, Inc Supplier No. 02100894.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aluminum silicate clay used as an emulsifier and adsorbent for proteins and virus particles.

  • Bentonit is used as Aluminum Silicate (clay), Emulsifier and adsorbent for proteins and virus particles

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1302-78-9
Molecular Formula Principally montmorillonite - Al2O3 . 4SiO2 . 2H2O
MDL Number MFCD00130611
Synonym Montmorillonite

Specifications

Color Gray
Linear Formula Principally montmorillonite - Al2O3 . 4SiO2 . 2H2O
Quantity 500 g
Grade Not Available
Physical Form Powder
Chemical Name or Material BENTONITE
Recommended Storage Store at Room Temperature (15-30°C), desiccate.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.