Learn More
Crescent Chemical Co Inc BENTONITE-SF RESEARCH GRADE
SDP
Supplier:  Crescent Chemical Co Inc SE14515.02
Encompass_Preferred
Bentonite-SF research grade, NF Emulsifier and adsorbent for proteins and viruses. Conforms to NF in its gel-forming capacity. For detoxification use a 2 % aqueous suspension. Suitable for the removal of cyanate in urea solutions. Size - 2.5KG Storage
Catalog No. 50-247-338