Learn More
Medchemexpress LLC Benz[a]anthracene-d12 | 1718-53-2 | 99.7% | 240.36 | 10 MG
SDP
Catalog No. 5000452824
Encompass_Preferred
Benz[a]anthracene-d12 is a deuterium labeled Benz[a]anthracene, intended for research use only. This compound is valuable as a tracer and as an internal standard for quantitative analysis methods.
  • Can be used as a tracer
  • Suitable for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only
Catalog No. 50-004-52824 Supplier Medchemexpress LLC Supplier No. HYW012278S10MG
Benz[a]anthracene-d12 is a deuterium labeled Benz[a]anthracene, intended for research use only. This compound is valuable as a tracer and as an internal standard for quantitative analysis methods.
  • Can be used as a tracer
  • Suitable for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only