Learn More
Medchemexpress LLC Benz[a]anthracene-d12 | 1718-53-2 | 99.74% | 240.36 | 5 MG
SDP
Catalog No. 5000453572
Encompass_Preferred
Benz[a]anthracene-d12 is the deuterium labeled Benz[a]anthracene. It is intended for research use only. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes, such as deuterium, have been incorporated into drug molecules for quantitation during drug development, and deuteration may impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeling may affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-53572 Supplier Medchemexpress LLC Supplier No. HYW012278S5MG
Benz[a]anthracene-d12 is the deuterium labeled Benz[a]anthracene. It is intended for research use only. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes, such as deuterium, have been incorporated into drug molecules for quantitation during drug development, and deuteration may impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeling may affect pharmacokinetic and metabolic profiles of drugs