Learn More
Medchemexpress LLC Benz[a]anthracene-d12 | 1718-53-2 | MFCD00020473 | 99.7% | 240.36 | 50 MG
SDP
Catalog No. 5000452243
Encompass_Preferred
This product is a deuterium-labeled compound, which means it contains heavy isotopes of hydrogen. These types of compounds are useful as tracers and internal standards for quantitative analysis in various techniques like NMR, GC-MS, or LC-MS. Deuteration can influence the pharmacokinetic and metabolic profiles of pharmaceuticals. It appears as a white to off-white solid with a purity of 99.74%.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-52243 Supplier Medchemexpress LLC Supplier No. HYW012278S50MG
This product is a deuterium-labeled compound, which means it contains heavy isotopes of hydrogen. These types of compounds are useful as tracers and internal standards for quantitative analysis in various techniques like NMR, GC-MS, or LC-MS. Deuteration can influence the pharmacokinetic and metabolic profiles of pharmaceuticals. It appears as a white to off-white solid with a purity of 99.74%.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.