Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benz[a]anthracene, MilliporeSigma™ Supelco™

Catalog No. 111020001 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0001 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0001 Supplier MilliporeSigma Supelco Supplier No. 48563
Add to Cart
Add to Cart

Benz[a]anthracene is a colorless plate like material, which belongs to the group of polycyclic aromatic hydrocarbon.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 56-55-3
Physical Form Neat
Linear Formula C18H12
MDL Number MFCD00003599
Melting Point 157°C to 159°C
Boiling Point 437.6°C
Synonym 1,2-Benzanthracene; Tetraphene
Type Food and Cosmetic Component Standard
Quantity 100 mg
Formula Weight 228.29
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.