Learn More
Medchemexpress LLC Benz[cd]indolium, inner salt (FD-1080) | 1151888-25-3 | MFCD34676913 | 96.3% | 743.3 g·mol⁻¹ | C40H39ClN2O6S2 | 5 MG
SDP
Catalog No. 5000262657
Encompass_Preferred
FD-1080 free acid is a near-infrared II (NIR-II) fluorophore with excitation at 1064 nm and emission at 1080 nm, provided as a brown to black solid for in vivo fluorescence imaging. It has high listed purity (~96.3%) and should be stored at -80°C, protected from light, and under nitrogen to maintain stability.
  • Excitation at 1064 nm and emission at 1080 nm for NIR-II imaging.
  • High purity suitable for imaging applications.
  • Brown to black solid form for convenient handling.
  • Stable under recommended storage conditions (-80°C, protected from light, under nitrogen).
  • Available in multiple small-scale sizes for preclinical use.
Catalog No. 50-002-62657 Supplier Medchemexpress LLC Supplier No. HY133852A5MG
Add to Cart
Add to Cart
FD-1080 free acid is a near-infrared II (NIR-II) fluorophore with excitation at 1064 nm and emission at 1080 nm, provided as a brown to black solid for in vivo fluorescence imaging. It has high listed purity (~96.3%) and should be stored at -80°C, protected from light, and under nitrogen to maintain stability.
  • Excitation at 1064 nm and emission at 1080 nm for NIR-II imaging.
  • High purity suitable for imaging applications.
  • Brown to black solid form for convenient handling.
  • Stable under recommended storage conditions (-80°C, protected from light, under nitrogen).
  • Available in multiple small-scale sizes for preclinical use.