Learn More
Genscript Corporation Benz-Neburase His
Catalog No. 5000056955
Encompass
This enzyme can be used to remove all forms of DNA and RNA including double stranded single stranded linearized and circular forms
Catalog No. 50-000-56955 Supplier Genscript Corporation Supplier No. Z03626500
This enzyme can be used to remove all forms of DNA and RNA including double stranded single stranded linearized and circular forms